Assay Details
Gene Symbol: Sec13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006978.1 | NP_001006979.1 | 9 | 1160 | 122 | |
GenBank mRNA | BC084705.1 | - | 9 | 1160 | 7301091 | 122 |
FQ212944.1 | - | 9 | 1180 | 122 | ||
FQ216715.1 | - | 9 | 1178 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
