Assay Details
Gene Symbol: Cables1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107404.1 | NP_001100874.1 | 1 | 448 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
