Assay Details
Gene Symbol: Usp38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751862.3 | XP_003751910.1 | 1 | 357 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003751862.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
