Assay Details
Gene Symbol: Serinc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY539942.1 | - | 1894 | 117 |
Gene Symbol: LOC501503 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | DQ100477.1 | - | 2 | 4296 | 117 |
Gene Symbol: Lrrgt00014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | U83119.1 | - | 1 | 2599 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
