Assay Details
Gene Symbol: Krt18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053976.1 | NP_446428.1 | 7 | 1284 | 77 | |
GenBank mRNA | BC091275.1 | - | 7 | 1284 | 7383311 | 77 |
U67992.1 | - | 1 | 84 | 77 |
Gene Symbol: LOC683212 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760823.1 | XP_008759045.1 | 2 | 1244 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
