Assay Details
Gene Symbol: Crp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY310153.1 | - | 1790 | 93 | ||
AY318961.1 | - | 1790 | 93 | |||
AY321318.1 | - | 1787 | 93 | |||
AY321324.1 | - | 1790 | 93 | |||
AY325196.1 | - | 1790 | 93 | |||
AY325218.1 | - | 1790 | 93 | |||
AY325232.1 | - | 1790 | 93 |
Gene Symbol: Lrrgt00014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | U83119.1 | - | 2 | 3706 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
