Assay Details
Gene Symbol: Fat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191705.1 | NP_001178634.1 | 12601 | 70 | ||
XM_003749273.3 | XP_003749321.2 | 844 | 70 | |||
XM_006232292.2 | XP_006232354.1 | 13692 | 70 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
