Assay Details
Gene Symbol: Bnipl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128187.2 | NP_001121659.1 | 10 | 1434 | 73 | |
XM_006232889.2 | XP_006232951.1 | 9 | 1773 | 73 | ||
GenBank mRNA | BC166747.1 | - | 10 | 1434 | 5598850 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
