Assay Details
Gene Symbol: Gipc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008761552.1 | XP_008759774.1 | 1 | 1213 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008761552.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
