Assay Details
Gene Symbol: Qsox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109898.1 | NP_001103368.1 | 12 | 2680 | 61 | |
NM_053431.3 | NP_445883.1 | 13 | 1829 | 61 | ||
GenBank mRNA | AF217799.1 | - | 13 | 1833 | 61 | |
AF285078.1 | - | 13 | 1851 | 61 | ||
AY623665.1 | - | 12 | 2735 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
