Assay Details
Gene Symbol: Tgfb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031131.1 | NP_112393.1 | 8 | 1789 | 111 | |
XM_006250448.2 | XP_006250510.1 | 7 | 2615 | 111 | ||
GenBank mRNA | AF153012.2 | - | 8 | 1789 | 111 | |
AF153013.1 | - | 7 | 1705 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
