Assay Details
Gene Symbol: Sec22b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025686.1 | NP_001020857.1 | 5 | 989 | 95 | |
GenBank mRNA | BC098725.1 | - | 5 | 989 | 7378284 | 95 |
FQ211503.1 | - | 1014 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
