Assay Details
Gene Symbol: Ect2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006227648.2 | XP_006227710.1 | 6 - 7 | 899 | 71 |
Gene Symbol: NEWGENE_2319083 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008758666.1 | XP_008756888.1 | 14 - 15 | 2091 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
