Assay Details
Gene Symbol: Cryz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012183.2 | NP_001012183.1 | 9 | 1815 | 132 | |
XM_006233520.2 | XP_006233582.1 | 10 | 2051 | 132 | ||
GenBank mRNA | BC078927.1 | - | 9 | 1817 | 7106704 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
