Assay Details
Gene Symbol: Kctd20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256148.1 | XP_006256210.1 | 8 | 3159 | 95 | |
XM_006256149.1 | XP_006256211.1 | 8 | 3150 | 95 | ||
XM_006256151.2 | XP_006256213.1 | 7 | 3071 | 95 | ||
XM_006256152.2 | XP_006256214.1 | 7 | 3032 | 95 | ||
XM_228042.6 | XP_228042.4 | 7 | 3368 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
