Assay Details
Gene Symbol: Ggt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019235.2 | NP_062108.2 | 1300 | 94 | ||
GenBank mRNA | BC087018.1 | - | 1300 | 7191011 | 94 | |
U76252.1 | - | 1037 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
