Assay Details
Gene Symbol: Slc16a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772974.1 | XP_008771196.1 | 6 | 5093 | 118 | |
XM_008772975.1 | XP_008771197.1 | 5 | 4948 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
