Assay Details
Gene Symbol: Cd24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012752.3 | NP_036884.1 | 2 | 1627 | 84 | |
GenBank mRNA | BC064439.1 | - | 2 | 1610 | 5623514 | 84 |
BC081851.1 | - | 2 | 1622 | 7110912 | 84 | |
CA509517.1 | - | 1 | 240 | 84 | ||
U49062.1 | - | 3 | 1549 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
