Assay Details
Gene Symbol: Cdk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007743.1 | NP_001007744.1 | 7 | 1053 | 86 | |
GenBank mRNA | BC082037.1 | - | 7 | 1053 | 7130973 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
