Assay Details
Gene Symbol: Adal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008762166.1 | XP_008760388.1 | 2 - 3 | 404 | 88 | |
XM_008762169.1 | XP_008760391.1 | 1 - 2 | 140 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
