Assay Details
Gene Symbol: Wtap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113543.1 | NP_001107015.1 | 6 | 1146 | 99 | |
NM_001113544.1 | NP_001107016.1 | 6 | 1080 | 99 | ||
GenBank mRNA | BQ202659.1 | - | 1 | 99 | ||
FQ228682.1 | - | 6 | 1185 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
