Assay Details
Gene Symbol: Cramp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006246051.2 | XP_006246113.1 | 18 - 19 | 4031 | 89 | |
XM_006246052.2 | XP_006246114.1 | 18 - 19 | 3695 | 89 | ||
XM_006246053.2 | XP_006246115.1 | 18 - 19 | 3481 | 89 | ||
XM_008767606.1 | XP_008765828.1 | 18 - 19 | 4028 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
