Assay Details
Gene Symbol: E2f5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760849.1 | XP_008759071.1 | 7 | 2199 | 160 | |
XM_574892.6 | XP_574892.1 | 8 | 1670 | 160 | ||
GenBank mRNA | U31668.1 | - | 8 | 1019 | 160 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -



Gene Ontology Categories:
