Assay Details
Gene Symbol: Zbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006235674.2 | XP_006235736.1 | 1 | 2336 | 108 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006235674.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
