Assay Details
Gene Symbol: Tnip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006236605.2 | XP_006236667.1 | 10 - 11 | 880 | 93 | |
XM_006236607.2 | XP_006236669.1 | 9 - 10 | 751 | 93 | ||
XM_008762999.1 | XP_008761221.1 | 9 - 10 | 832 | 93 | ||
XM_008763000.1 | XP_008761222.1 | 10 - 11 | 845 | 93 | ||
XM_008763001.1 | XP_008761223.1 | 8 - 9 | 820 | 93 | ||
XM_008763002.1 | XP_008761224.1 | 9 - 10 | 736 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
