Assay Details
Gene Symbol: Shc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164060.1 | NP_001157532.1 | 9 | 1395 | 65 | |
NM_053517.2 | NP_445969.2 | 10 | 962 | 65 | ||
GenBank mRNA | BC088298.1 | - | 10 | 962 | 7323045 | 65 |
CB813924.1 | - | 3 | 242 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
