Assay Details
Gene Symbol: H1fx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_575600.6 | XP_575600.1 | 1 | 1894 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_575600.6 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
