Assay Details
Gene Symbol: Crp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY310153.1 | - | 677 | 99 | ||
AY318961.1 | - | 677 | 99 | |||
AY321318.1 | - | 674 | 99 | |||
AY321324.1 | - | 677 | 99 | |||
AY325196.1 | - | 677 | 99 | |||
AY325218.1 | - | 677 | 99 | |||
AY325232.1 | - | 677 | 99 |
Gene Symbol: LOC501503 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | DQ100477.1 | - | 2 | 4290 | 99 |
Gene Symbol: Lrrgt00014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | U83119.1 | - | 1 | 2593 | 99 |
Gene Symbol: Serinc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY539942.1 | - | 1888 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
