Assay Details
Gene Symbol: Nr4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008763702.1 | XP_008761924.1 | 1 | 1099 | 70 | |
XM_008763703.1 | XP_008761925.1 | 1 | 1099 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008763702.1, XM_008763703.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
