Assay Details
Gene Symbol: B4galt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107965.1 | NP_001101435.1 | 7 | 1494 | 103 | |
XM_006238667.2 | XP_006238729.1 | 7 | 1564 | 103 | ||
GenBank mRNA | FQ214130.1 | - | 5 | 985 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
