Assay Details
Gene Symbol: Twf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008521.1 | NP_001008521.1 | 9 | 1705 | 74 | |
GenBank mRNA | BC086536.1 | - | 9 | 1705 | 7106678 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
