Assay Details
Gene Symbol: LOC103694910 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773907.1 | XP_008772129.1 | 210 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008773907.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
