Assay Details
Gene Symbol: Syt14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751296.3 | XP_003751344.1 | 5 - 6 | 937 | 106 | |
XM_003751297.3 | XP_003751345.1 | 5 - 6 | 989 | 106 | ||
XM_008769895.1 | XP_008768117.1 | 4 - 5 | 1972 | 106 | ||
XM_008769896.1 | XP_008768118.1 | 4 - 5 | 1972 | 106 | ||
XM_008769897.1 | XP_008768119.1 | 4 - 5 | 1972 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
