Assay Details
Gene Symbol: Mpv17l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764515.1 | XP_008762737.1 | 1 | 559 | 81 | |
XM_008764516.1 | XP_008762738.1 | 1 | 558 | 81 | ||
XM_008764517.1 | XP_008762739.1 | 1 | 560 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008764515.1, XM_008764516.1, XM_008764517.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
