Assay Details
Gene Symbol: Dcaf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100718.1 | NP_001094188.1 | 9 | 1988 | 62 | |
XM_006240279.2 | XP_006240341.1 | 9 | 1790 | 62 | ||
XM_008764782.1 | XP_008763004.1 | 9 | 1843 | 62 | ||
GenBank mRNA | BC079428.1 | - | 4 | 1107 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
