Assay Details
Gene Symbol: Catsperb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764913.1 | XP_008763135.1 | 2 - 3 | 124 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
