Assay Details
Gene Symbol: Rnf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008770271.1 | XP_008768493.1 | 8 | 2811 | 125 | |
XM_008770272.1 | XP_008768494.1 | 8 | 2766 | 125 | ||
XM_008770273.1 | XP_008768495.1 | 8 | 2899 | 125 | ||
GenBank mRNA | FQ211855.1 | - | 7 | 2661 | 125 | |
FQ223606.1 | - | 8 | 2795 | 125 | ||
FQ225378.1 | - | 1 | 1408 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
