Assay Details
Gene Symbol: Pacsin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008765660.1 | XP_008763882.1 | 1 | 4494 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008765660.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
