Assay Details
Gene Symbol: Rpl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC063811.1 | - | 4 - 5 | 494 | 6919340 | 115 |
BC081801.1 | - | 4 - 5 | 449 | 7192390 | 115 | |
FQ209448.1 | - | 3 - 4 | 332 | 115 | ||
FQ209532.1 | - | 3 - 4 | 332 | 115 | ||
FQ209571.1 | - | 3 - 4 | 332 | 115 | ||
FQ212557.1 | - | 3 - 4 | 294 | 115 | ||
FQ212566.1 | - | 3 - 4 | 332 | 115 | ||
FQ212656.1 | - | 3 - 4 | 332 | 115 | ||
FQ212896.1 | - | 3 - 4 | 332 | 115 | ||
FQ212979.1 | - | 1 - 2 | 124 | 115 | ||
FQ213045.1 | - | 3 - 4 | 332 | 115 | ||
FQ213454.1 | - | 4 - 5 | 483 | 115 | ||
FQ215017.1 | - | 3 - 4 | 288 | 115 | ||
FQ215042.1 | - | 3 - 4 | 332 | 115 | ||
FQ215264.1 | - | 3 - 4 | 332 | 115 | ||
FQ215283.1 | - | 3 - 4 | 332 | 115 | ||
FQ215533.1 | - | 3 - 4 | 332 | 115 | ||
FQ215586.1 | - | 3 - 4 | 332 | 115 | ||
FQ215649.1 | - | 3 - 4 | 332 | 115 | ||
FQ216342.1 | - | 3 - 4 | 332 | 115 | ||
FQ216573.1 | - | 3 - 4 | 332 | 115 | ||
FQ216754.1 | - | 3 - 4 | 332 | 115 | ||
FQ216792.1 | - | 3 - 4 | 332 | 115 | ||
FQ218195.1 | - | 3 - 4 | 332 | 115 | ||
FQ219327.1 | - | 3 - 4 | 332 | 115 | ||
FQ219561.1 | - | 3 - 4 | 332 | 115 | ||
FQ219701.1 | - | 3 - 4 | 332 | 115 | ||
FQ219716.1 | - | 3 - 4 | 332 | 115 | ||
FQ220231.1 | - | 3 - 4 | 330 | 115 | ||
FQ220328.1 | - | 1 - 2 | 154 | 115 | ||
FQ221130.1 | - | 3 - 4 | 294 | 115 | ||
FQ221674.1 | - | 3 - 4 | 332 | 115 | ||
FQ226216.1 | - | 3 - 4 | 332 | 115 | ||
FQ228709.1 | - | 3 - 4 | 332 | 115 | ||
FQ228734.1 | - | 4 - 5 | 483 | 115 | ||
FQ228797.1 | - | 3 - 4 | 332 | 115 | ||
FQ228847.1 | - | 3 - 4 | 332 | 115 | ||
FQ228910.1 | - | 3 - 4 | 332 | 115 | ||
FQ228929.1 | - | 3 - 4 | 332 | 115 | ||
FQ228930.1 | - | 3 - 4 | 332 | 115 | ||
FQ229086.1 | - | 3 - 4 | 332 | 115 | ||
FQ229116.1 | - | 3 - 4 | 332 | 115 | ||
FQ229723.1 | - | 1 - 2 | 72 | 115 | ||
FQ229849.1 | - | 3 - 4 | 291 | 115 | ||
FQ231653.1 | - | 3 - 4 | 332 | 115 | ||
FQ232105.1 | - | 3 - 4 | 332 | 115 | ||
FQ232562.1 | - | 3 - 4 | 332 | 115 | ||
FQ232771.1 | - | 3 - 4 | 332 | 115 | ||
FQ233898.1 | - | 3 - 4 | 332 | 115 | ||
FQ234293.1 | - | 3 - 4 | 332 | 115 | ||
FQ234521.1 | - | 3 - 4 | 332 | 115 | ||
FQ234619.1 | - | 3 - 4 | 332 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
