Assay Details
Gene Symbol: Igf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241191.2 | XP_006241253.1 | 6 | 3243 | 96 | |
XM_006241192.1 | XP_006241254.1 | 6 | 3157 | 96 | ||
XM_006241193.2 | XP_006241255.1 | 5 | 3190 | 96 | ||
XM_006241194.2 | XP_006241256.1 | 5 | 3281 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
