Assay Details
Gene Symbol: Chst15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006230430.2 | XP_006230492.1 | 8 | 5561 | 69 | |
XM_006230431.2 | XP_006230493.1 | 8 | 6301 | 69 | ||
XM_006230433.2 | XP_006230495.1 | 8 | 5796 | 69 | ||
XM_008759868.1 | XP_008758090.1 | 8 | 4557 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
