Assay Details
Gene Symbol: Clpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_217313.8 | XP_217313.6 | 1 | 171 | 105 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_217313.8 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
