Assay Details
Gene Symbol: Hpse | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022605.1 | NP_072127.1 | 12 | 2540 | 99 | |
GenBank mRNA | AF184967.1 | - | 12 | 2540 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
