Assay Details
Gene Symbol: Dtx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107157.1 | NP_001100627.1 | 12 | 2585 | 104 | |
XM_006249158.1 | XP_006249220.1 | 11 | 2531 | 104 | ||
XM_006249159.1 | XP_006249221.1 | 10 | 2393 | 104 | ||
XM_008769137.1 | XP_008767359.1 | 11 | 2386 | 104 | ||
GenBank mRNA | AY724523.1 | - | 9 | 2228 | 104 | |
BC166738.1 | - | 11 | 2472 | 7937292 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
