Assay Details
Gene Symbol: Pank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006231276.2 | XP_006231338.1 | 7 | 9549 | 122 | |
XM_006231277.2 | XP_006231339.1 | 6 | 9377 | 122 | ||
XM_006231278.2 | XP_006231340.1 | 8 | 6897 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
