Assay Details
Gene Symbol: Prkar1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013181.1 | NP_037313.1 | 11 | 1300 | 82 | |
XM_008768311.1 | XP_008766533.1 | 11 | 1356 | 82 | ||
GenBank mRNA | M17086.1 | - | 11 | 1300 | 82 | |
U75932.1 | - | 8 | 814 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
