Assay Details
Gene Symbol: Gabre | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF255385.1 | - | 2890 | 109 | ||
AF255612.1 | - | 1972 | 109 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
