Assay Details
Gene Symbol: Kdm6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773695.1 | XP_008771917.1 | 7 - 8 | 563 | 96 | |
XM_008773696.1 | XP_008771918.1 | 6 - 7 | 518 | 96 | ||
GenBank mRNA | FJ775728.1 | - | 419 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
