Assay Details
Gene Symbol: Rccd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229455.2 | XP_006229517.1 | 6 - 7 | 1676 | 87 | |
XM_006229456.2 | XP_006229518.1 | 6 - 7 | 1332 | 87 | ||
XM_218819.7 | XP_218819.5 | 6 - 7 | 1577 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
