Assay Details
Gene Symbol: Arhgap21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191693.1 | NP_001178622.1 | 6550 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
